Please login to your online account to display your discounted pricing

anti-SNX27, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human SNX27 (within residues 150-200). [Swiss-Prot No. Q96L92]
Antigen SNX27
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP145282SS

 
novus biologicals canada
NBP145282SS
Unlabeled 0.025mL Each for $109.00

NBP145282

 
novus biologicals canada
NBP145282
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human SNX27 (within residues 150-200). [Swiss-Prot No. Q96L92]
Antigen SNX27
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Human and mouse. Immunogen sequence has 94% identity to rat and bovine.
Gene Accession No. Q96L92
Gene ID (Entrez) 81609
Formulation PBS, 30% glycerol
Concentration 1mg/mL