Please login to your online account to display your discounted pricing

anti-SNX2, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 350-400 of human Snx2 was used as the immunogen.
Antigen SNX2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224513

 
novus biologicals canada
NBP224513
Unlabeled 0.1mg Each for $257.41

NBP224513SS

 
novus biologicals canada
NBP224513SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 350-400 of human Snx2 was used as the immunogen.
Antigen SNX2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, horse, platypus, cow, rat, rhesus monkey, mouse, chicken and chimpanzee and 94% homologous in xenopus.
Gene ID (Entrez) 6643
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL