missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
SNF8, ESCRT-II complex subunit, homolog (S. cerevisiae), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human SNF8 protein.
Supplier: Abnova Corporation H00011267B01
Description
SNF8, VPS25 (MIM 610907), and VPS36 (MIM 610903) form ESCRT-II (endosomal sorting complex required for transport II), a complex involved in endocytosis of ubiquitinated membrane proteins. SNF8, VPS25, and VPS36 are also associated in a multiprotein complex with RNA polymerase II elongation factor (ELL; MIM 600284) (Slagsvold et al., 2005 [PubMed 15755741]; Kamura et al., 2001 [PubMed 11278625]).[supplied by OMIM
Sequence: MHRRGVGAGAIAKKKLAEAKYKERGTVLAEDQLAQMSKQLDMFKTNLEEFASKHKQEIRKNPEFRVQFQDMCATIGVDPLASGKGFWSEMLGVGDFYYELGVQIIEVCLALKHRNGGLITLEELHQQVLKGRGKFAQDVSQDDLIRAIKKLKALGTGFGIIPVGGTYLIQSVPAELNMDHTVVLQLAEKNGYVTVSEIKASLKWETERARQVLEHLLKEGLAWLDLQAPGEAHYWLPALFTDLYSQEITAEEAREALPSpecifications
SNF8, ESCRT-II complex subunit, homolog (S. cerevisiae) | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human SNF8 protein. | |
SNF8 | |
Dot3/EAP30/VPS22 | |
Mouse | |
50 μL | |
Transcription Regulation | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
NM_007241.2 | |
SNF8 | |
SNF8 (NP_009172.2, 1 a.a. ∼ 258 a.a) full-length human protein. | |
RUO | |
Yes | |
11267 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |