Please login to your online account to display your discounted pricing

anti-Snail, Polyclonal, Novus Biologicals™

$109.00 - $259.02

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human SNAIL protein (between residues 200-300) [UniProt O95863]
Antigen Snail
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP229626

 
novus biologicals canada
NBP229626
Unlabeled 0.1mg Each for $259.02

NBP229626SS

 
novus biologicals canada
NBP229626SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human SNAIL protein (between residues 200-300) [UniProt O95863]
Antigen Snail
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Human, mouse and monkey. Predicted to react with rat, mink, feline and hamster based on 100% sequence homology.
Gene ID (Entrez) 6615
Formulation PBS
Concentration 1.0mg/mL