Please login to your online account to display your discounted pricing

anti-Snail, Polyclonal, Novus Biologicals™

$139.00 - $362.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 80-130 of human SNAI1 was used as the immunogen
Antigen Snail
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227184

 
novus biologicals canada
NBP227184
Unlabeled 0.1mg Each for $362.00

NBP227184SS

 
novus biologicals canada
NBP227184SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 80-130 of human SNAI1 was used as the immunogen
Antigen Snail
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, horse and dog, and 94% homologous mouse and rat.
Gene Accession No. O95863
Gene ID (Entrez) 6615
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL