Please login to your online account to display your discounted pricing

anti-Smoothened, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 190-240 of human SMO was used as the immunogen for this antibody.
Antigen Smoothened
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224543

 
novus biologicals canada
NBP224543
Unlabeled 0.1mg Each for $329.00

NBP224543SS

 
novus biologicals canada
NBP224543SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 190-240 of human SMO was used as the immunogen for this antibody.
Antigen Smoothened
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human ( isoforms CRA_a, CRA_b, CRA_c), rhesus monkey and chimpanzee, 94% homologous in horse, cow and dog, 88% homologous in rat, mouse, chicken, opossum, zebrafish and Xenopus and 83% homologous in pufferfish.
Gene ID (Entrez) 6608
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL