Please login to your online account to display your discounted pricing

anti-Smoothened, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224543SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224543SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 6608
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Concentration 0.5mg/mL
Immunogen A portion of amino acids 190-240 of human SMO was used as the immunogen for this antibody.
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human ( isoforms CRA_a, CRA_b, CRA_c), rhesus monkey and chimpanzee, 94% homologous in horse, cow and dog, 88% homologous in rat, mouse, chicken, opossum, zebrafish and Xenopus and 83% homologous in pufferfish.
Conjugate Unlabeled
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.025mg
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Format Affinity Purified
Regulatory Status RUO
Antigen Smoothened