Please login to your online account to display your discounted pricing

anti-Smoothened, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224543

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224543

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 6608
Primary or Secondary Primary
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human ( isoforms CRA_a, CRA_b, CRA_c), rhesus monkey and chimpanzee, 94% homologous in horse, cow and dog, 88% homologous in rat, mouse, chicken, opossum, zebrafish and Xenopus and 83% homologous in pufferfish.
Concentration 0.5mg/mL
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Conjugate Unlabeled
Immunogen A portion of amino acids 190-240 of human SMO was used as the immunogen for this antibody.
Format Affinity Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Quantity 0.1mg
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Regulatory Status RUO
Antigen Smoothened