missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-SMC4, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding amino acid residues 47-61 of human SMC4. Genbank accession number: BAA73535.
Antigen SMC4
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056395 Novus Biologicals Canada
NB10056395
0.1mg Each for $257.41
NB10056395T Novus Biologicals Canada
NB10056395SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
SMC4
Purified
RUO
The band corresponding to the apparent molecular weight of human SMC4 is blocked by immunizing peptide.
10051
0.5mg/mL
A synthetic peptide corresponding amino acid residues 47-61 of human SMC4. Genbank accession number: BAA73535.
Polyclonal
Primary
Western Blot
NP_001099000
PBS containing 0.2% gelatin