Please login to your online account to display your discounted pricing

anti-SMC4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056395SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056395T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 10051
Formulation PBS containing 0.2% gelatin
Monoclonal or Polyclonal Polyclonal
Concentration 0.5mg/mL
Quantity 0.025mg
Regulatory Status RUO
Immunogen A synthetic peptide corresponding amino acid residues 47-61 of human SMC4. Genbank accession number: BAA73535.
Cross Reactivity Human
Gene Accession No. NP_001099000
Primary or Secondary Primary
Conjugate Unlabeled
Antigen SMC4
Host Species Rabbit
Test Specificity The band corresponding to the apparent molecular weight of human SMC4 is blocked by immunizing peptide.
Format Purified
Applications Western Blotting