Please login to your online account to display your discounted pricing

anti-SMC4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056395

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056395

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 10051
Test Specificity The band corresponding to the apparent molecular weight of human SMC4 is blocked by immunizing peptide.
Formulation PBS containing 0.2% gelatin
Gene Accession No. NP_001099000
Conjugate Unlabeled
Format Purified
Applications Western Blotting
Immunogen A synthetic peptide corresponding amino acid residues 47-61 of human SMC4. Genbank accession number: BAA73535.
Quantity 0.1mg
Antigen SMC4
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Regulatory Status RUO
Concentration 0.5mg/mL
Cross Reactivity Human