Please login to your online account to display your discounted pricing

anti-Smallpox A27L, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Between amino acids 10 to 60 of the A27L protein were used as the immunogen.
Antigen Smallpox A27L
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056456

 
novus biologicals canada
NB10056456
Unlabeled 0.1mg Each for $255.80

NB10056456T

 
novus biologicals canada
NB10056456SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Between amino acids 10 to 60 of the A27L protein were used as the immunogen.
Antigen Smallpox A27L
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen has 100% identity in human, camel, cow, and rabbit. Cross reacts with Variola virus.
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL