Please login to your online account to display your discounted pricing

anti-Smad7, Polyclonal, Novus Biologicals™

$192.00 - $406.00

Specifications

Host Species Rabbit
Immunogen This antibody was raised against amino acids 12-29 (RLWRSRAPGGEDEEEGAG) and amino acids 36-50 (ELRGEGATDSRAHGA) of human SMAD7. This sequence is 100% or highly conserved between human and multiple species.
Antigen Smad7
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056688

 
novus biologicals canada
NB10056688
Unlabeled 0.1mg Each for $406.00

NB10056688T

 
novus biologicals canada
NB10056688SS
Unlabeled 0.025mg Each for $192.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was raised against amino acids 12-29 (RLWRSRAPGGEDEEEGAG) and amino acids 36-50 (ELRGEGATDSRAHGA) of human SMAD7. This sequence is 100% or highly conserved between human and multiple species.
Antigen Smad7
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity In A549, an approx. 45 kDa band is observed. This may cross-react with SMAD6 depending on cell or tissue-specific expression. Cross reacts with Human, Mouse and Rat. Predicted to react with Chimpanzee, Dog, Pufferfish, Zebrafish and Xenopus.
Gene Accession No. NP_005895
Gene ID (Entrez) 4092
Formulation PBS (Filter sterilized)
Concentration 0.5mg/mL