Please login to your online account to display your discounted pricing

anti-Smad7, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056688SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056688T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 4092
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Immunogen This antibody was raised against amino acids 12-29 (RLWRSRAPGGEDEEEGAG) and amino acids 36-50 (ELRGEGATDSRAHGA) of human SMAD7. This sequence is 100% or highly conserved between human and multiple species.
Quantity 0.025mg
Formulation PBS (Filter sterilized)
Concentration 0.5mg/mL
Test Specificity In A549, an approx. 45 kDa band is observed. This may cross-react with SMAD6 depending on cell or tissue-specific expression. Cross reacts with Human, Mouse and Rat. Predicted to react with Chimpanzee, Dog, Pufferfish, Zebrafish and Xenopus.
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Antigen Smad7
Format Purified
Conjugate Unlabeled
Primary or Secondary Primary
Gene Accession No. NP_005895