Please login to your online account to display your discounted pricing

anti-Smad4, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Isotype IgG
Immunogen This antibody was developed against synthetic peptides corresponding to amino acids 186-223 and amino acids 509-544 of human SMAD4 (GenBank Accession No. NP_005350.1). These peptide sequences are specific to SMAD4
Antigen Smad4
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224951

 
novus biologicals canada
NBP224951
Unlabeled 0.1mg Each for $255.80

NBP224951SS

 
novus biologicals canada
NBP224951SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen This antibody was developed against synthetic peptides corresponding to amino acids 186-223 and amino acids 509-544 of human SMAD4 (GenBank Accession No. NP_005350.1). These peptide sequences are specific to SMAD4
Antigen Smad4
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene Accession No. Q13485
Gene ID (Entrez) 4089
Formulation 100μg antibody in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL