Please login to your online account to display your discounted pricing

anti-Smad2, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Amino acids 234-249 (DQQLNQSMDTGSPAEL) of human SMAD2 protein was used as the immunogen.
Antigen Smad2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056462

 
novus biologicals canada
NB10056462
Unlabeled 0.1mg Each for $329.00

NB10056462T

 
novus biologicals canada
NB10056462SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Amino acids 234-249 (DQQLNQSMDTGSPAEL) of human SMAD2 protein was used as the immunogen.
Antigen Smad2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The SMAD2 amino acid sequence used as immunogen is 100% homologous in human (isoform 1: 467 amino acids; isoform 2: 437 amino acids), mouse, rat, horse, cow, chicken and Xenopus. Reacts with Bovine, Chicken, Chimpanzee, Dog, Human, Mouse and Rat.
Gene Accession No. NP_005892
Gene ID (Entrez) 4087
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL