Please login to your online account to display your discounted pricing

anti-Smad2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056462

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056462

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 4087
Primary or Secondary Primary
Formulation PBS containing 0.05% BSA
Conjugate Unlabeled
Gene Accession No. NP_005892
Regulatory Status RUO
Test Specificity The SMAD2 amino acid sequence used as immunogen is 100% homologous in human (isoform 1: 467 amino acids; isoform 2: 437 amino acids), mouse, rat, horse, cow, chicken and Xenopus. Reacts with Bovine, Chicken, Chimpanzee, Dog, Human, Mouse and Rat.
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Antigen Smad2
Host Species Rabbit
Format Purified
Immunogen Amino acids 234-249 (DQQLNQSMDTGSPAEL) of human SMAD2 protein was used as the immunogen.
Quantity 0.1mg