Please login to your online account to display your discounted pricing

anti-SMAC/Diablo, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056164

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056164

  • / Each
 

Description & Specifications

Specifications

Antigen SMAC/Diablo
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Format Antisera
Formulation PBS
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 56-73 (AVPIAQKSEPHSLSSEAL) of Smac/DIABLO was used as immunogen
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 56616
Test Specificity Smac .
Target Species Human