Please login to your online account to display your discounted pricing

anti-SMAC/Diablo, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056391SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056391T

  • / Each
 

Description & Specifications

Specifications

Antigen SMAC/Diablo
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS containing 0.2% gelatin
Gene Accession No. Q9NR28
Host Species Rabbit
Immunogen This rabbit polyclonal antibody was developed against a synthetic peptide corresponding to amino acids 226 to 239 of human Smac (Genbank accession no. AAF87716).
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 56616
Target Species Human