Please login to your online account to display your discounted pricing

anti-SMAC/Diablo, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056391

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056391

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 56616
Concentration 0.5mg/mL
Quantity 0.1mg
Gene Accession No. Q9NR28
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Cross Reactivity Human
Monoclonal or Polyclonal Polyclonal
Formulation PBS containing 0.2% gelatin
Primary or Secondary Primary
Immunogen This rabbit polyclonal antibody was developed against a synthetic peptide corresponding to amino acids 226 to 239 of human Smac (Genbank accession no. AAF87716).
Format Purified
Antigen SMAC/Diablo
Host Species Rabbit