Please login to your online account to display your discounted pricing

anti-SMAC/Diablo, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056166SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056166T

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 56616
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Antiserum
Antigen SMAC/Diablo
Formulation PBS
Test Specificity Smac . Cross reacts with Gerbil, Human, Mouse and Rat.
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Immunogen Full-length recombinant human SMAC was used as immunogen.
Regulatory Status RUO
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Quantity 0.025mL