missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-SMAC/Diablo, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This rabbit polyclonal antibody was developed against a synthetic peptide corresponding to amino acids 226 to 239 of human Smac (Genbank accession no. AAF87716).
Antigen SMAC/Diablo
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056391 Novus Biologicals Canada
NB10056391
0.1mg Each for $255.80
NB10056391T Novus Biologicals Canada
NB10056391SS
0.025mg Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
SMAC/Diablo
Purified
RUO
Q9NR28
PBS containing 0.2% gelatin
This rabbit polyclonal antibody was developed against a synthetic peptide corresponding to amino acids 226 to 239 of human Smac (Genbank accession no. AAF87716).
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
56616
0.5mg/mL