Please login to your online account to display your discounted pricing

anti-SMAC/Diablo, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen This rabbit polyclonal antibody was developed against two synthetic peptides corresponding to amino acids 199 to 212 and amino acids 226 to 239 of human Smac (Genbank accession no. AAF87716).
Antigen SMAC/Diablo
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056311

 
novus biologicals canada
NB10056311
Unlabeled 0.1mg Each for $259.02

NB10056311T

 
novus biologicals canada
NB10056311SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This rabbit polyclonal antibody was developed against two synthetic peptides corresponding to amino acids 199 to 212 and amino acids 226 to 239 of human Smac (Genbank accession no. AAF87716).
Antigen SMAC/Diablo
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Smac .
Gene Accession No. Q9NR28
Gene ID (Entrez) 56616
Formulation PBS
Concentration 0.5mg/mL