Please login to your online account to display your discounted pricing

anti-SMAC/Diablo, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056166

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056166

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 56616
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.05mL
Regulatory Status RUO
Conjugate Unlabeled
Formulation PBS
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Test Specificity Smac . Cross reacts with Gerbil, Human, Mouse and Rat.
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Antiserum
Immunogen Full-length recombinant human SMAC was used as immunogen.
Antigen SMAC/Diablo