Please login to your online account to display your discounted pricing

anti-SMAC/Diablo, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056164SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056164T

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Smac .
Gene ID (Entrez) 56616
Immunogen A synthetic peptide corresponding to amino acids 56-73 (AVPIAQKSEPHSLSSEAL) of Smac/DIABLO was used as immunogen
Formulation PBS
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Host Species Rabbit
Format Antiserum
Regulatory Status RUO
Quantity 0.025mL
Antigen SMAC/Diablo