Please login to your online account to display your discounted pricing

anti-SLITRK1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP149541

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP149541

  • / Each
 

Description & Specifications

Specifications

Antigen SLITRK1
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blot
Concentration 0.65mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. Q96PX8
Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human Slitrk1 protein (within residues 250-400). [Swiss-Prot Q96PX8]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 114798
Test Specificity Human and mouse. Immunogen sequence has 84% homology to rat.
Target Species Human
Target Species Murine