Please login to your online account to display your discounted pricing

anti-SLIRP, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of human SLIRP protein. [Entrez-Prot No. AAX58600]
Antigen SLIRP
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11037258T

 
novus biologicals canada
NB11037258SS
Unlabeled 0.025mL Each for $139.00

NB11037258

 
novus biologicals canada
NB11037258
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of human SLIRP protein. [Entrez-Prot No. AAX58600]
Antigen SLIRP
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Gene Accession No. Q9GZT3
Gene ID (Entrez) 81892
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL