Please login to your online account to display your discounted pricing

anti-SLC28A3, Polyclonal, Novus Biologicals™

$109.00 - $307.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of human SLC28A3 (between amino acids 2-60) [UniProt Q9HAS3]
Antigen SLC28A3
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP233301

 
novus biologicals canada
NBP233301
Unlabeled 0.1mg Each for $307.00

NBP233301SS

 
novus biologicals canada
NBP233301SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of human SLC28A3 (between amino acids 2-60) [UniProt Q9HAS3]
Antigen SLC28A3
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene Accession No. Q9HAS3
Gene ID (Entrez) 64078
Formulation PBS
Concentration 1.0mg/mL