Please login to your online account to display your discounted pricing

anti-SLC28A3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP233301SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP233301SS

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Monoclonal or Polyclonal Polyclonal
Concentration 1.0mg/mL
Host Species Rabbit
Quantity 0.025mg
Antigen SLC28A3
Format Purified
Conjugate Unlabeled
Gene Accession No. Q9HAS3
Gene ID (Entrez) 64078
Applications Western Blotting
Primary or Secondary Primary
Immunogen Synthetic peptide made to an N-terminal portion of human SLC28A3 (between amino acids 2-60) [UniProt Q9HAS3]
Regulatory Status RUO
Cross Reactivity Human