Please login to your online account to display your discounted pricing

anti-SLC28A3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP233301

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP233301

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Formulation PBS
Cross Reactivity Human
Host Species Rabbit
Antigen SLC28A3
Gene Accession No. Q9HAS3
Concentration 1.0mg/mL
Gene ID (Entrez) 64078
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Format Purified
Quantity 0.1mg
Immunogen Synthetic peptide made to an N-terminal portion of human SLC28A3 (between amino acids 2-60) [UniProt Q9HAS3]