Please login to your online account to display your discounted pricing

anti-Slap, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 175-225 of human SLAP was used as the immunogen for the antibody.
Antigen Slap
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224696SS

 
novus biologicals canada
NBP224696SS
Unlabeled 0.025mg Each for $109.00

NBP224696

 
novus biologicals canada
NBP224696
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 175-225 of human SLAP was used as the immunogen for the antibody.
Antigen Slap
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Test Specificity The UNC5CL amino acid sequence used as immunogen for the antibody is 100% homologous in human and chimpanzee and 88% homologous in rhesus monkey.
Gene ID (Entrez) 6503
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL