Please login to your online account to display your discounted pricing

anti-Slap, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224696SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224696SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Test Specificity The UNC5CL amino acid sequence used as immunogen for the antibody is 100% homologous in human and chimpanzee and 88% homologous in rhesus monkey.
Regulatory Status RUO
Concentration 0.5mg/mL
Antigen Slap
Quantity 0.025mg
Gene ID (Entrez) 6503
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Primary or Secondary Primary
Host Species Rabbit
Applications Immunohistochemistry (Paraffin)
Format Purified
Cross Reactivity Human
Immunogen A portion of amino acids 175-225 of human SLAP was used as the immunogen for the antibody.