Please login to your online account to display your discounted pricing

anti-SIRT7, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056364

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056364

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Test Specificity There is no corresponding 40 kD band seen in mouse or rat liver cell lysate.
Conjugate Unlabeled
Formulation PBS containing 0.05% BSA
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 51547
Concentration 0.5mg/mL
Antigen SIRT7
Format Purified
Gene Accession No. Q9NRC8
Host Species Rabbit
Immunogen A mixture of synthetic peptides corresponding to amino acids 35-51 and 361-377 of human SIRT7 was used as immunogen.
Quantity 0.1mg
Regulatory Status RUO
Cross Reactivity Human