Please login to your online account to display your discounted pricing

anti-SIRT7, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11081762

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11081762

  • / Each
 

Description & Specifications

Specifications

Concentration 1mg/mL
Primary or Secondary Primary
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Host Species Rabbit
Format Affinity Purified
Antigen SIRT7
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 51547
Conjugate Unlabeled
Gene Accession No. Q9NRC8
Formulation Tris-citrate/phosphate, pH 7-8
Immunogen Synthetic peptide made to an internal portion of human SIRT7 (within residues 50-150). [Swiss-Prot No. Q9NRC8]
Quantity 0.1mL
Regulatory Status RUO