Please login to your online account to display your discounted pricing

anti-SIRT7, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11081746SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11081746T

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Conjugate Unlabeled
Gene Accession No. Q9NRC8
Quantity 0.025mL
Host Species Rabbit
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen SIRT7
Gene ID (Entrez) 51547
Test Specificity Human, mouse, bovine and rat. Immunogen sequence has 93% homology to porcine.
Formulation Tris-citrate/phosphate, pH 7-8
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Concentration 1mg/mL
Regulatory Status RUO
Format Affinity Purified
Immunogen Synthetic peptide made to an internal portion of human SIRT7 (within residues 200-300). [Swiss-Prot No. Q9NRC8]