Please login to your online account to display your discounted pricing

anti-SIRT7, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11081746

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11081746

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Formulation Tris-citrate/phosphate, pH 7-8
Test Specificity Human, mouse, bovine and rat. Immunogen sequence has 93% homology to porcine.
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Concentration 1mg/mL
Gene ID (Entrez) 51547
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Format Affinity Purified
Gene Accession No. Q9NRC8
Regulatory Status RUO
Immunogen Synthetic peptide made to an internal portion of human SIRT7 (within residues 200-300). [Swiss-Prot No. Q9NRC8]
Antigen SIRT7
Quantity 0.1mL
Host Species Rabbit