Please login to your online account to display your discounted pricing

anti-SIRT7, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11081744SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11081744T

  • / Each
 

Description & Specifications

Specifications

Antigen SIRT7
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. Q9NRC8
Host Species Rabbit
Primary or Secondary Primary
Gene ID (Entrez) 51547
Test Specificity Human and mouse. Immunogen sequence has 88% homology to bovine and porcine and 82% homology to Drosophila.
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Immunogen Synthetic peptide made to an internal portion of human SIRT7 (within residues 100-200). [Swiss-Prot No. Q9NRC8]
Quantity 0.025mL
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Format Affinity Purified