Please login to your online account to display your discounted pricing

anti-SIRT7, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human SIRT7 (within residues 50-150). [Swiss-Prot No. Q9NRC8]
Antigen SIRT7
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11081762

 
novus biologicals canada
NB11081762
Unlabeled 0.1mL Each for $309.05

NB11081762T

 
novus biologicals canada
NB11081762SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human SIRT7 (within residues 50-150). [Swiss-Prot No. Q9NRC8]
Antigen SIRT7
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Gene Accession No. Q9NRC8
Gene ID (Entrez) 51547
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL