Please login to your online account to display your discounted pricing

anti-SIRT7, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056364SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056364T

  • / Each
 

Description & Specifications

Specifications

Antigen SIRT7
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS containing 0.05% BSA
Gene Accession No. Q9NRC8
Host Species Rabbit
Immunogen A mixture of synthetic peptides corresponding to amino acids 35-51 and 361-377 of human SIRT7 was used as immunogen.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 51547
Test Specificity There is no corresponding 40 kD band seen in mouse or rat liver cell lysate.
Target Species Human