Please login to your online account to display your discounted pricing

anti-SIRT6, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the mouse SIRT6 protein (within residues 250-334). [Swiss-Prot P59941]
Antigen SIRT6
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1002522SS

 
novus biologicals canada
NB1002522SS
Unlabeled 0.025mL Each for $139.00

NB1002522

 
novus biologicals canada
NB1002522
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the mouse SIRT6 protein (within residues 250-334). [Swiss-Prot P59941]
Antigen SIRT6
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Mouse. Rat reactivity reported in scientific literature (PMID: 25288139) Immunogen sequence has 78% homology to human.
Gene ID (Entrez) 51548
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1.0mg/mL