Please login to your online account to display your discounted pricing

anti-SIRT6, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002522SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002522SS

  • / Each
 

Description & Specifications

Specifications

Antigen SIRT6
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the mouse SIRT6 protein (within residues 250-334). [Swiss-Prot P59941]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 51548
Test Specificity Mouse. Rat reactivity reported in scientific literature (PMID: 25288139) Immunogen sequence has 78% homology to human.
Target Species Murine
Target Species Rat