Please login to your online account to display your discounted pricing

anti-SIN3A, Polyclonal, Novus Biologicals™

$160.00 - $362.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the mouse Sin3A protein (between residues 1-50) [UniProt Q60520]
Antigen SIN3A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB6001263

 
novus biologicals canada
NB6001263
Unlabeled 0.1mL Each for $362.00

NB6001263SS

 
novus biologicals canada
NB6001263SS
Unlabeled 0.025mL Each for $160.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the mouse Sin3A protein (between residues 1-50) [UniProt Q60520]
Antigen SIN3A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity mSin 3A. Cross-reacts with Human and Mouse. Not yet tested in other species.
Gene Accession No. Q96ST3
Gene ID (Entrez) 25942
Formulation PBS
Concentration 1mg/mL