Please login to your online account to display your discounted pricing

anti-SIN3A, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6001263

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6001263

  • / Each
 

Description & Specifications

Specifications

Antigen SIN3A
Applications Immunoprecipitation
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS
Gene Accession No. Q96ST3
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the mouse Sin3A protein (between residues 1-50) [UniProt Q60520]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 25942
Test Specificity mSin 3A. Cross-reacts with Human and Mouse. Not yet tested in other species.
Target Species Human
Target Species Murine