Please login to your online account to display your discounted pricing

anti-Sin1/MAPKAP1, Polyclonal, Novus Biologicals™

$139.00 - $395.29

Specifications

Host Species Rabbit
Isotype IgG
Immunogen The immunogen recognized by this antibody maps to a region between residue 470 and the C-terminus (residue 522) of human stress-activated map kinase interacting protein 1 using the numbering given in entry NP_001006618.1 (GeneID 79109).
Antigen Sin1/MAPKAP1
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056482T

 
novus biologicals canada
NB10056482SS
Unlabeled 0.025mg Each for $139.00

NB11040424

 
novus biologicals canada
NB11040424
Unlabeled 0.1mg Each for $395.29
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen The immunogen recognized by this antibody maps to a region between residue 470 and the C-terminus (residue 522) of human stress-activated map kinase interacting protein 1 using the numbering given in entry NP_001006618.1 (GeneID 79109).
Antigen Sin1/MAPKAP1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Proximity Ligation Assay
Applications Western Blotting
Test Specificity Human, Mouse. Based on 100% sequence identity, this antibody is predicted to react with Rat, Chicken, Turkey, Sheep, Bovine, Dog, Horse, Guinea pig, Pig, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, West Indian ocean coelacanth, Duckbill platypus, Tasmanian devil and Zebra finch.
Gene Accession No. Q9BPZ7
Gene ID (Entrez) 79109
Formulation Tris-citrate/phosphate buffer, pH 7 to 8
Concentration 1.0mg/mL