Please login to your online account to display your discounted pricing

anti-Siglec-9, APC, Clone: 191240, R&D Systems


Manufacturer: r & d systems  RDSFAB1139A025

 View more versions of this product

Catalog No. FAB1139A025

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Antigen Siglec-9
Quantity 25 Tests
Host Species Mouse
Isotype IgG2a
Conjugate APC
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Immunogen Mouse myeloma cell line NS0-derived recombinant human Siglec-9. Gln18-Gly348 (predicted)
Cross Reactivity Human
Format Purified
Clone 191240
Purification Method Protein A or G purified from hybridoma culture supernatant