Please login to your online account to display your discounted pricing

anti-Siglec-9, APC, Clone: 191240, R&D Systems

$155.12 - $441.90

Specifications

Antigen Siglec-9
Clone 191240
Cross Reactivity Human
Format Purified
Host Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB1139A025

 
r & d systems
RDSFAB1139A025
APC 25 Tests Each for $155.12

FAB1139A100

 
r & d systems
RDSFAB1139A100
APC 100 Tests Each for $441.90
Description & Specifications

Specifications

Antigen Siglec-9
Clone 191240
Cross Reactivity Human
Format Purified
Host Species Mouse
Regulatory Status RUO
Immunogen Mouse myeloma cell line NS0-derived recombinant human Siglec-9. Gln18-Gly348 (predicted)
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Flow Cytometry