Please login to your online account to display your discounted pricing

anti-Siglec-9, APC, Clone: 191240, R&D Systems


Manufacturer: r & d systems  RDSFAB1139A100

 View more versions of this product

Catalog No. FAB1139A100

  • / Each
 

Description & Specifications

Specifications

Antigen Siglec-9
Clone 191240
Conjugate Allophycocyanin
Cross Reactivity Human
Format Purified
Host Species Murine
Immunogen Mouse myeloma cell line NS0-derived recombinant human Siglec-9. Gln18-Gly348 (predicted)
Isotype IgG2a
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human