Please login to your online account to display your discounted pricing

anti-Siglec-3/CD33, Fluorescein, Clone: 6C5/2, R&D Systems


Manufacturer: r & d systems  RDSFAB1137F025

 View more versions of this product

Catalog No. FAB1137F025

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Format Purified
Isotype IgG1
Antigen Siglec-3/CD33
Host Species Mouse
Cross Reactivity Human
Quantity 25 Tests
Clone 6C5/2
Primary or Secondary Primary
Conjugate Fluorescein
Immunogen Recombinant human Siglec-3/CD33. Extracellular domain
Purification Method Protein A or G purified from hybridoma culture supernatant
Monoclonal or Polyclonal Monoclonal