Please login to your online account to display your discounted pricing

anti-Siglec-3/CD33, Fluorescein, Clone: 6C5/2, R&D Systems

$116.01 - $272.44

Specifications

Antigen Siglec-3/CD33
Clone 6C5/2
Cross Reactivity Human
Format Purified
Host Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB1137F100

 
r & d systems
RDSFAB1137F100
Fluorescein 100 Tests Each for $272.44

FAB1137F025

 
r & d systems
RDSFAB1137F025
Fluorescein 25 Tests Each for $116.01
Description & Specifications

Specifications

Antigen Siglec-3/CD33
Clone 6C5/2
Cross Reactivity Human
Format Purified
Host Species Mouse
Regulatory Status RUO
Immunogen Recombinant human Siglec-3/CD33. Extracellular domain
Isotype IgG1
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Applications:

Flow Cytometry