Please login to your online account to display your discounted pricing

anti-Siglec-3/CD33, APC, Clone: 6C5/2, R&D Systems


Manufacturer: r & d systems  RDSFAB1137A025

 View more versions of this product

Catalog No. FAB1137A025

  • / Each
 

Description & Specifications

Specifications

Antigen Siglec-3/CD33
Clone 6C5/2
Conjugate Allophycocyanin
Cross Reactivity Human
Format Purified
Host Species Murine
Immunogen Recombinant human Siglec-3/CD33. Extracellular domain
Isotype IgG1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human